Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
Product Code. 10683622
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
Material:
Duran Glass
Duran Tubing, PTFE
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material unitSize
10683622 100 mL Duran Glass Each
10000491 50 mL Duran Glass Each
10244531 50 mL Duran Tubing, PTFE Each
10468212 100 mL Duran Tubing, PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10683622 Supplier Lenz Laborglasintrumente Supplier No. 05305014
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Receivers with straight adapter
  • With cooling jacket, graduated, with socket and cone NS 29/32
  • Vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Capacity (Metric) 100 mL
Standard Taper Joint Socket and Cone NS 29/32