Learn More
Excel Healthcare Ltd™ Distiller Cleaner
Efficient cleaning solution
Brand:  Excel Healthcare Ltd™ 288
Additional Details : Weight : 5.00000kg
Product Code. 16320132
  • £26.55 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Distiller Cleaning Enhances The Removal Of Debris Prior To Sterilisation
  • It Is Vital That The Water And Detergent Are Well Maintained, Cleaned And Changed At Regular Intervals To Ensure Maximum Cleaning Effectiveness
  • Htm 01-05 Recommends That Intruments Placed In Ultrasonic Baskets Must Be Fully Immersed In The Cleaning Solution
  • After The Process Has Taken Place, Instruments Must Be Thoroughly Cleaned And Rinsed Using Distilled Water In Order To Remove Residual Soil And Detergents With Minimum Risk Of Salt Deposition
Specifications
Specifications
Distiller Cleaner
12 oz.
340 g