Learn More
Gibco™ Distilled Water
Product Code. 11518886 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
Unit Size:
1 L
100mL
5000mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11518886 500 mL 500mL
11508886 100 mL 100mL
10114292 1000 mL 1 L
11528886 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 11518886 Supplier Gibco™ Supplier No. 15230089
  • ONLINE EXCLUSIVE PRICE
    £18.95
    £17.65 / 500mL
    Save £1.30
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Research Use Only. All usage must comply with product instructions.
Distilled Water
Gibco™ Distilled Water is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, Gibco™ Distilled Water features:
  • Cell culture quality
  • Proven performance
  • Use in cell culture and molecular biology applications
    Cell culture quality
    Gibco™ Distilled Water is prepared by distillation and filtration (0.1μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.
    Proven performance
    The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.
    Use in cell culture and molecular biology applications
    Gibco™ Distilled Water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.
    Product Use
    For Research Use Only. Not for use in diagnostic procedures.
    Dual-site cGMP Manufacturing and Quality System
    Gibco™ Distilled Water is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Distilled Water product made in our Grand Island facility (15230-170). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description Distilled Water, 500mL, Bottle(s), Membrane-Filtered Purification, Sterile, 15 to 30°C Storage Conditions, Room Temperature/Ambient, Fewer Resources Used/Less Waste, ISO 13485/cGMP/FDA Certified
    Sterility Sterile
    Treatment(s) Not DEPC-treated
    Certificate of Analysis ISO 13485, cGMP, FDA
    Quantity 500 mL