missing translation for 'onlineSavingsMsg'
Learn More

BOLA Distillation Apparata

Product Code. 10319231
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
4000 mL
6000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10319231 500 mL Each
10104491 1000 mL Each
10247451 2000 mL Each
10787302 4000 mL Each
10629601 6000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10319231 Supplier BOLA Supplier No. B28003
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for the distillation of strong alkaline or acid products as well as very aggressive solvents.

  • Distillation Apparata of PTFE - with SAFE LAB-System consisting of: Reaction vessel
  • flat flange (B281-03) Gasket
  • flat flange (B 282-02) Lid
  • flat flange (B 283-02) Joining piece
  • flat flange (B 284-02) Dropping funnel
  • 125 ml (B 285-01) Supporting device for thermometer (B 286-03)
  • Thermometer for flasks (B 287-03)
  • Stirrer bearing (B288-02)
  • Moon-Shaped stirrer shaft (C 376-12)
  • Stillhead 2x NS 29/32 (B 289-03)
  • Distillation thermometer (B 290-03)
  • Liebig condenser 270 mm (B 291-02)
  • Receiver adapter (B 292-02)
  • Vacuum stopcock (B 293-02)
  • Round-bottom flask (A 158-06)

Specifications

Capacity (Metric) 500 mL
Dimensions (L x W x H) 450 x 600 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.