Learn More
Corning™ Dissolvable Microcarriers
GREENER_CHOICE
Product Code. 15823895 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
100 g
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15823895 10 g Each
15813895 5 g Each
15803895 1 g Each
15833895 100 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15823895 Supplier Corning™ Supplier No. 4989
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning™ Dissolvable Microcarriers are designed to simplify cell harvest, separation and concentration processes and are ideal for bioprocess scale-up.
  • Offer cell separations that are faster, gentler, and more convenient than other standard commercially available microcarriers
  • Comprised of polygalacturonic acid (PGA) polymer chains cross linked via calcium ions pre-coated with either denatured collagen or Corning Synthemax™ II to enhance cell attachment for optimal yield and viability
  • Dissolvable within 10 to 20 minutes using EDTA and pectinase for easy separation of cells from the actual microcarriers
  • Narrow size distribution (1.02-1.03 g/cm3 density, 200-300μm fully hydrated) and large surface area (5000cm2/g) result in a more homogenous culture
  • Optical clarity for visualizing cell morphology
  • Presterilized to Sterility Assurance Level (SAL) of 10-6
  • Nonpyrogenic
  • Applications areas include cell therapy, vaccine production or large-volume adherent cell culture scale-up and cell banking
TRUSTED_SUSTAINABILITY
Specifications
Description Glass vial
Culture Area 5000 cm2/g
For Use With (Application) Large-volume adherent cell culture scale-up and cell banking
Surface Treatment Corning™ Synthemax II
Product Type Dissolvable Microcarrier
Material Polygalacturonic acid (PGA) polymer chains cross linked via calcium ions
Particle Size (Metric) 200 to 300 μm fully hydrated
Quantity 10 g