missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Disposable Transfer Pipets

Product Code. 13496408
Change view
Click to view available options
Volume (Metric):
5 mL
Unit Size:
Pack of 4000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13496408 5 mL Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 13496408 Supplier Simport™ Supplier No. P2005610S
Estimated Shipment: 04-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Eliminate the hazard of broken glass and exposure to infectious materials with these all-in-one pipets. Simport™ Disposable Transfer Pipets are molded from see-through low density polyethylene and inert to biological fluids and most acids.

  • LDPE transfer pipets
  • Put an end to matching rubber bulbs with glass pipets
  • Can be sealed and refrigerated
  • Work well whenever there is a need for quick, safe transfer of fluids
  • Temperature resistant down to -196 °C. Can be gaz (EtO) sterilized
  • Choose between 12 popular models, available in several sizes, tip designs and lengths, in sterile or non-sterile packaging
  • Available with graduations

Specifications

Bulb Draw (Metric) 1.9 mL
Diameter (Metric) Outer 6.7 mm
Material LDPE
Drops/mL 25
For Use With (Application) For quick, safe transfer of fluids
Graduated Yes
Quantity 10 Pack
Volume (Metric) 5 mL
Length (Metric) 15.5 cm
Type Transfer Pipet
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.