missing translation for 'onlineSavingsMsg'
Learn More

Buerkle Disposable Powder Sampler PharmaDispo
GREENER_CHOICEBIOPHARMA

Product Code. 30245471 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.03 mL (0.01 mL per chamber)
0.15 mL (0.05 mL per chamber)
0.3 mL (0.1 mL per chamber)
1.5 mL (0.5 mL per chamber)
3 mL (1 mL per chamber)
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 30245471

Brand: Buerkle 53938000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A disposable multi-target sampler that takes three separate target point samples with a single sampling. It is manufactured, assembled and individually packaged under class 7 (ISO) clean room conditions.

The three separate sampling chambers of the PharmaDispo allow simultaneous sampling from different layers or zones. This allows the material composition to be investigated in a differentiated manner across multiple layers, which is particularly beneficial when a single sample does not sufficiently characterise the material. The target point samples taken can be analysed individually or combined to form a representative composite sample.

An advanced ergonomic design allows the sampler to sit comfortably in the hand and sampling can be performed with one hand. A sliding mechanism with an end stop makes it easy to open and close the sampler.

The PharmaDispo sampler accurately and safely takes small powder samples for quality control from open containers.

  • Take small individual samples from different layers
  • Easy to use: Sliding mechanism with end stop for user-friendly operation
  • Clean room manufactured, individually packaged and optionally sterilized by gamma rays
TRUSTED_SUSTAINABILITY

Specifications

Product Type Powder Sampler
Material Polystyrene
Autoclavable Not Autoclavable
Capacity (Metric) 0.03 mL (0.01 mL per chamber)
Disposable Single-use
Sterility Non-sterile
No. of Chambers 3
Packaging Type Pack of 20 pieces
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.