missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Disposable Magnetic Stirring Bars

Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.

Brand:  Bel-Art™ SP Scienceware™ F37112-0016

151.55 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : 03

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11754146

  • £167.00 / Pack of 100
Estimated Shipment: 05-07-2024
to see stock.
Add to basket

Description

Description

  • Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Specifications

Specifications

8 mm
Mixing radioactive reagents when cross contamination might be a concern
100/Pk.
Yes
41.3 mm
41.3 x 8mm
White
Alnico Magnet
ISO 9001:2008
Polygon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers