Learn More
Brand™ Blaubrand™ IntraMARK™ Disposable Micropipets
Constructed of disposable glass and conforms to DIN ISO 7550 standards. BRAND™ Blaubrand™ IntraMARK™ Disposable Micropipets include ring mark, and are CE-marked according to IVD Directive 98/79 CE. All surfaces are fire polished for smoothness and to minimize pipet adapter wear. Color-coded to ISO for clear identification.
Brand:  Brand™ 708718
Code : NEW
Additional Details : Weight : 0.60300kg
 View more versions of this product
Product Code. 15202599
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • With ring mark
  • Conform to DIN ISO 7550
  • Calibrated to contain (TC, In.)
  • CE-marked according to IVD Directive 98/79 CE.
  • Fire polished surfaces inside and out are particulary smooth
  • The aspiration end is also fire polished to minimize pipet adapter wear.
  • Color code to ISO for clear identification.
Specifications
Specifications
≤±0.25%
DIN ISO 7550, IVD-Directive 98/79 CE-marked
Glass
Black
Single-use
To Contain
Marks at 20μL
250/Box
125 mm