missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 48 g/m2
Shop All Medicom Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 60
Description
- Light and breathable polypropylene material
- Loose shape allows user to maintain good movement
- Collarless shirt
- Pants with elasticated waist
- Individual packaging for hygiene and controlled distribution
- 48 g/m2 fabric weight
Tekniske data
Tekniske data
| Size | Small |
| Material | Polypropylene |
| Color | Navy |
| Sleeve Style | Short Sleeve |
| Disposable | Single-use |
| For Use With (Application) | Dental, Medical |
| No. per Pack | 60 Pk. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion