missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 40 g/m2

Product Code. p-7227164 Shop All Medicom Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 17700179

Brand: Kolmi Hopen™ J40800/S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

These sets of disposable tunics and pants provide extra protection for healthcare workers.

  • Light and breathable polypropylene material
  • Loose shape allows user to maintain good movement
  • Distribution of fibers guarantees opacity
  • Collarless shirt
  • Pants with elasticated waist
  • Individual packaging for hygiene and controlled distribution
  • 40 g/m2 fabric weight

Specifications

Size Small
Material SMS
Color Blue
Sleeve Style Short Sleeve
Disposable Single-use
For Use With (Application) Dental, Medical
No. per Pack 50 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.