missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Disposable Gloves Zero Waste Box™

Product Code. 17885925
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
17885925 Small
17865835 Medium
17875835 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17885925 Supplier Terracycle™ Supplier No. CS45SUK
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Disposable Gloves using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Vinyl, nitrile, and latex gloves

Approximate waste units accepted:

  • Small: 600 gloves
  • Medium: 1700 gloves
  • Large: 3100 gloves

Do not send in:

  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Autoclaved material
  • Gloves that are contaminated with materials considered hazardous or dangerous
  • Heavy duty, fabric, and leather gloves
  • Hairnets, earplugs, and lab coats or garments
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which present a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass

Specifications

Capacity (Metric) 7 kg
Capacity (English) 15 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 25 x 25 x 46 cm
Disposable Single-use
Latex Free Yes
Size Small
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.