missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Disposable Gloves and Face Masks Zero Waste Box™
GREENER_CHOICE

Product Code. p-7249609
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17825905

Brand: Terracycle™ CS176SUK

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Recycle Disposable Gloves and Face Masks using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Disposable protective face masks such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment)
  • Face shields and visors
  • Disposable vinyl, nitrile and latex gloves

Approximate waste units accepted:

  • Small: 520 units
  • Medium: 1400 units
  • Large: 2600 units

Do not send in:

  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, care homes, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
    • Autoclaved material
    • Gloves that are contaminated with materials considered hazardous or dangerous
    • Fabric, homemade or paper masks or other types of protective garments
    • Hairnets, earplugs, and lab coats or garments
    • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which presents a danger to the environment, or to people.

Specifications

Capacity (Metric) 7 kg
Capacity (English) 15 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 25 x 25 x 46 cm
Disposable Single-use
Latex Free Yes
Size Small
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.