missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Disposable Gloves and Face Masks Zero Waste Box™

Product Code. 17835905
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17835905 Large Each
17825905 Small Each
17815815 Medium -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17835905 Supplier Terracycle™ Supplier No. CS176LUK
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Disposable Gloves and Face Masks using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Disposable protective face masks such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment)
  • Face shields and visors
  • Disposable vinyl, nitrile and latex gloves

Approximate waste units accepted:

  • Small: 520 units
  • Medium: 1400 units
  • Large: 2600 units

Do not send in:

  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, care homes, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
    • Autoclaved material
    • Gloves that are contaminated with materials considered hazardous or dangerous
    • Fabric, homemade or paper masks or other types of protective garments
    • Hairnets, earplugs, and lab coats or garments
    • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which presents a danger to the environment, or to people.

Specifications

Capacity (Metric) 25 kg
Capacity (English) 55 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Latex Free Yes
Size Large
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.