missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Disposable Gloves and Face Masks Zero Waste Box™

Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
Use this Zero Waste Box to recycle:
- Disposable protective face masks such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment)
- Face shields and visors
- Disposable vinyl, nitrile and latex gloves
Approximate waste units accepted:
- Small: 520 units
- Medium: 1400 units
- Large: 2600 units
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, care homes, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Autoclaved material
- Gloves that are contaminated with materials considered hazardous or dangerous
- Fabric, homemade or paper masks or other types of protective garments
- Hairnets, earplugs, and lab coats or garments
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which presents a danger to the environment, or to people.
Specifications
Specifications
| Capacity (Metric) | 15 kg |
| Capacity (English) | 33 lb. |
| Color | Beige, Green, Red, White |
| Dimensions (L x W x H) | 30.5 x 30.5 x 84 cm |
| Disposable | Single-use |
| Latex Free | Yes |
| Size | Medium |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Cardboard, Plastic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction