missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Disposable Garments Zero Waste Box™
Recycle Disposable Garments using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Brand: Terracycle™ CS67-M-UK
This item is not returnable.
View return policy
Description
Use this Zero Waste Box to recycle:
- Any brand and size of disposable garments, overalls, chemical protection suits, smocks and disposable shoe covers
Approximate waste units accepted:
- Small: 15 garments
- Medium: 35 garments
- Large: 65 garments
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Autoclaved material
- Protective masks
- Eyewear
- Respirators and hard hats
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which present a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass
Specifications
15 kg | |
Beige, Green, Red, White | |
Single-use | |
Medium | |
Not Autoclavable | |
Recycling Box |
33 lb. | |
30.5 x 30.5 x 84 cm | |
Yes | |
Non-sterile | |
Cardboard, Plastic |