missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Disposable Funnel for Powders, LaboPlast™/SteriPlast™
GREENER_CHOICEBIOPHARMA

Product Code. 17520846 Shop All Bürkle Products
Click to view available options
Color:
Blue
White
Material:
Bio-PE
Polystyrene
Polystyrene, Detectable
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17520846

Brand: Buerkle™ 53786005

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Disposable Funnel for Powders, LaboPlast™/SteriPlast™ has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates, and suspensions, as well as other liquids.

  • For easy and reliable transferring
  • Wide opening prevents clogging
  • Suitable for use in laboratories and clean rooms or for sampling
  • Clean room manufactured (clean room class 7)
  • Individually packaged and optionally sterilized
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric)Top 100 mm
Diameter (Metric) Stem 25 mm
Material Polystyrene
Color White
For Use With (Application) Transferring Powders, Granulates and Suspensions
Sterility Non-sterile
Product Type Filling Funnel (Powders)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.