missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS Display Board Panel
Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-24 Classic
Unit Size:
Each
Description
This display board panel is a spare part for FastPrep-24 Classic.
Specifications
Specifications
| Product Type | Display Board Panel |
| For Use With (Equipment) | FastPrep-24 Classic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction