Learn More
IKA™ Dispersers, DIS-300-S
Product Code. 17217519 Shop All IKA Products
Click to view available options
Packaging Quantity:
10 Tubes
50 Tubes
Unit Size:
Pack of 10
Pack of 50
Product Code. 17217519
Brand: IKA™ 0020026959
Only null left
Add to basket
Add to basket
These Dispersers, DIS-300-S are disposable tubes equipped with a stainless steel dissolver blade. Compatible with ULTRA-TURRAX™ Tube Drive P control.
  • The lid of the DIS-300-S tube is bigger than the lid of the DIS-300-S-M tube and has no pierceable membrane
  • The bigger lid allows easy and convenient addition of solid samples
  • Features a stainless steel dissolver blade
  • Compatible with ULTRA-TURRAX™ Tube Drive P control
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube with Dissolver Blade
Capacity (Metric) 300 mL
Autoclavable Not Autoclavable
Blade Material Stainless Steel
For Use With (Application) Crushing, mixing, dissolving
For Use With (Equipment) ULTRA-TURRAX™ Tube Drive Power Control
Packaging Quantity 10 Tubes