missing translation for 'onlineSavingsMsg'
Learn More

Disperse Yellow 3, TRC

Product Code. 17902535
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17902535 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17902535 Supplier Toronto Research Chemicals Supplier No. TRCD493495500MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2832-40-8
Molecular Formula C15 H15 N3 O2
Quantity 500 mg
Synonym Disperse Yellow 3, Acetamide, N-[4-[2-(2-hydroxy-5-methylphenyl)diazenyl]phenyl]- (ACI), Acetamide, N-[4-[(2-hydroxy-5-methylphenyl)azo]phenyl]- (9CI), Acetanilide, 4'-(6-hydroxy-m-tolylazo)- (6CI), C.I. Disperse Yellow 3 (7CI, 8CI), N-[4-[2-(2-Hydroxy-5-methylphenyl)diazenyl]phenyl]acetamide (ACI), 4-Acetamido-2'-hydroxy-5'-methylazobenzene, 4'-[(2-Hydroxy-5-methylphenyl)azo]acetanilide, Acetamine Yellow CG, Acetate Fast Yellow G, Acetoquinone Light Yellow 4JLZ, Akasperse Yellow G, Amacel Yellow G, Artisil Yellow 2GN, Atrisil Direct Yellow G, C.I. 11855, C.I. Solvent Yellow 77, C.I. Solvent Yellow 92, C.I. Solvent Yellow 99, Calcosyn Yellow GC, Celliton Discharge Yellow GL, Celliton Fast Yellow G, Celliton Fast Yellow GA-CF, Celliton Yellow G, Celutate Yellow GH, Cibacet Yellow 2GC, Cibacet Yellow 2GC150, Cibacet Yellow GBA, Cibacete Yellow GBA, Cilla Fast Yellow G, Diacelliton Fast Yellow G, Disperse Yellow 3, Disperse Yellow G, Disperse Yellow Z, Dispersol Fast Yellow G, Dispersol Printing Yellow G, Dispersol Yellow A-G, DTNW 11, Durgacet Yellow G, Eastone Yellow GN, Esteroquinone Light Yellow 4JL, Fenacet Fast Yellow G, Interchem Acetate Yellow G, Interchem Hisperse Yellow GH, Intrasperse Yellow GBA, Intrasperse Yellow GBA Extra, Kayalon Fast Yellow G, Kayaset Yellow G, KCA Acetate Fast Yellow G, Kiwalon Polyester Fast Yellow G, Lurafix Yellow 142, Microsetile Yellow GR, Miketon Fast Yellow G, Nacelan Fast Yellow CG, Novalon Yellow 2GN, Nyloquinone Yellow 4J, Oplas Yellow 130, Ostacet Yellow P 2G, P 2G, Palanil Yellow G, Perliton Yellow G, Reliton Yellow C, Resiren Yellow TG, Safaritone yellow G, Serinyl Hosiery Yellow GD, Seriplas Yellow GD, Serisol Fast Yellow GD, Setacyl Yellow 2GN, Setacyl Yellow G, Setacyl Yellow P 2GL, Silotras Yellow TSG, Sumiplast Yellow FC, Supracet Fast Yellow G, Synten Yellow 2G, Terasil Yellow 2GC, Terasil Yellow GBA Extra, Transetile Yellow P-GR, Tulasteron Fast Yellow 2G-B, Vonteryl Yellow G, Waxoline Yellow RP-FW, Yellow Reliton G, Yellow Z
InChI Formula InChI=1S/C15H15N3O2/c1-10-3-8-15(20)14(9-10)18-17-13-6-4-12(5-7-13)16-11(2)19/h3-9,20H,1-2H3,(H,16,19)
SMILES CC(=O)Nc1ccc(cc1)N=Nc2cc(C)ccc2O
IUPAC Name N-[4-[(E)-(2-hydroxy-5-methylphenyl)diazenyl]phenyl]acetamide
Molecular Weight (g/mol) 269.3
Formula Weight 269.1164
Chemical Name or Material Disperse Yellow 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.