missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disperse Yellow 1 (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Disperse Yellow 1 |
| CAS | 119-15-3 |
| Molecular Formula | C12 H9 N3 O5 |
| Quantity | 5 g |
| Synonym | Phenol, p-(2,4-dinitroanilino)- (6CI,7CI,8CI), 4-[(2,4-Dinitrophenyl)amino]phenol, 2,4-Dinitro-4'-hydroxydiphenylamine, 4-(2,4-Dinitroanilino)phenol, 4-Hydroxy-2',4'-dinitrodiphenylamine, 4'-Hydroxy-2,4-dinitrodiphenylamine, Acetamine Yellow 2R, Acetoquinone Light Yellow 2RZ, Amacel Yellow RR, C.I. 10345, C.I. Disperse Yellow 1, C.I. Solvent Yellow 52, Celliton Fast Yellow RR, Celliton Fast Yellow RRA-CF, Cilla Fast Yellow RR, Disperse Fast Yellow 2K, Disperse Yellow 1, Disperse Yellow 2G, Disperse Yellow R, Disperse Yellow Stable 2K, Dispersol Fast Yellow A, Dispersol Printing Yellow A, Dispersol Yellow B-A, Fast Disperse Yellow 2K, Fenacet Fast Yellow 2R, Kayalon Fast Yellow RR, Microsetile Yellow 2R, N-(2,4-Dinitrophenyl)-4-aminophenol, NSC 13965, NSC 408078, NSC 6335, Nyloquinone Yellow 2R, Perliton Yellow RR, Permanent Yellow 2K, Reliton Yellow R, SRA Golden Yellow VIII, Serisol Fast Yellow A, Setacyl Yellow P-BS, Supracet Fast Yellow 2R, Supracet Yellow RR, Synten Yellow P 2R, Tulasteron Fast Yellow R-B |
| InChI Formula | InChI=1S/C12H9N3O5/c16-10-4-1-8(2-5-10)13-11-6-3-9(14(17)18)7-12(11)15(19)20/h1-7,13,16H |
| SMILES | Oc1ccc(Nc2ccc(cc2[N+](=O)[O-])[N+](=O)[O-])cc1 |
| IUPAC Name | 4-(2,4-dinitroanilino)phenol |
| Molecular Weight (g/mol) | 275.22 |
| Formula Weight | 275.0542 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?