missing translation for 'onlineSavingsMsg'
Learn More

Disperse Yellow 1 (Technical Grade), TRC

Product Code. 30383733
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383733 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383733 Supplier Toronto Research Chemicals Supplier No. TRCD4953231G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Yellow 1
CAS 119-15-3
Molecular Formula C12 H9 N3 O5
Quantity 1 g
Synonym Phenol, p-(2,4-dinitroanilino)- (6CI,7CI,8CI), 4-[(2,4-Dinitrophenyl)amino]phenol, 2,4-Dinitro-4'-hydroxydiphenylamine, 4-(2,4-Dinitroanilino)phenol, 4-Hydroxy-2',4'-dinitrodiphenylamine, 4'-Hydroxy-2,4-dinitrodiphenylamine, Acetamine Yellow 2R, Acetoquinone Light Yellow 2RZ, Amacel Yellow RR, C.I. 10345, C.I. Disperse Yellow 1, C.I. Solvent Yellow 52, Celliton Fast Yellow RR, Celliton Fast Yellow RRA-CF, Cilla Fast Yellow RR, Disperse Fast Yellow 2K, Disperse Yellow 1, Disperse Yellow 2G, Disperse Yellow R, Disperse Yellow Stable 2K, Dispersol Fast Yellow A, Dispersol Printing Yellow A, Dispersol Yellow B-A, Fast Disperse Yellow 2K, Fenacet Fast Yellow 2R, Kayalon Fast Yellow RR, Microsetile Yellow 2R, N-(2,4-Dinitrophenyl)-4-aminophenol, NSC 13965, NSC 408078, NSC 6335, Nyloquinone Yellow 2R, Perliton Yellow RR, Permanent Yellow 2K, Reliton Yellow R, SRA Golden Yellow VIII, Serisol Fast Yellow A, Setacyl Yellow P-BS, Supracet Fast Yellow 2R, Supracet Yellow RR, Synten Yellow P 2R, Tulasteron Fast Yellow R-B
InChI Formula InChI=1S/C12H9N3O5/c16-10-4-1-8(2-5-10)13-11-6-3-9(14(17)18)7-12(11)15(19)20/h1-7,13,16H
SMILES Oc1ccc(Nc2ccc(cc2[N+](=O)[O-])[N+](=O)[O-])cc1
IUPAC Name 4-(2,4-dinitroanilino)phenol
Molecular Weight (g/mol) 275.22
Formula Weight 275.0542
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.