missing translation for 'onlineSavingsMsg'
Learn More

Disperse Red 1 (Technical Grade), TRC

Product Code. 30378441
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30378441

Brand: Toronto Research Chemicals TRCD4734755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Red 1
CAS 2872-52-8
Molecular Formula C16 H18 N4 O3
Quantity 5 g
Synonym C.I. Disperse Red 1 (7CI,8CI), Celliton Scarlet B (6CI), Ethanol, 2-[ethyl[4-[(4-nitrophenyl)azo]phenyl]amino]- (9CI), 2-[Ethyl[4-[2-(4-nitrophenyl)diazenyl]phenyl]amino]ethanol, 1-[N-Ethyl-N-(2-hydroxyethyl)amino]-4-(4-nitrophenylazo)benzene, 4-Nitro-4'-[N-ethyl-N-(β-hydroxyethyl)amino]azobenzene, 4-Nitro-4'-[ethyl(2-hydroxyethyl)amino]azobenzene, 4-[(2-Hydroxyethyl)ethylamino]-4'-nitroazobenzene, 4-[4-(Nitrophenyl)azo]-N-ethyl-N-(2-hydroxyethyl)aniline, 4-[Ethyl(2-hydroxyethyl)amino]-4'-nitroazobenzene, 4-[N-(2-Hydroxyethyl)-N-ethylamino]-4'-nitroazobenzene, 4-[N-Ethyl-N-(2-hydroxyethyl)amino]-4'-nitroazobenzene, Acetamine Scarlet B, Acetate Fast Scarlet B, Acetoquinone Light Scarlet BLZ, Amacel Scarlet GB, C.I. 11110, C.I. Solvent Red 14, Calcosyn Brilliant Scarlet BN, Celliton Discharge Scarlet B, Celliton Scarlet BA-CF, Celutate Scarlet BH, Cibacet Scarlet 2B, Cibacet Scarlet BRN, Cibacet Scarlet BS, Cilla Scarlet B, D 3284, Diacelliton Fast Scarlet B, Diacelliton Scarlet B, Disperse Red 1, Disperse Red 1-glycerol-TDI copolymer, Disperse Red Zh, Disperse Scarlet B, Disperse Scarlet Zh, Dispersive ruby Zh, Dispersol Fast Scarlet B, Dispersol Scarlet B, Durgacet Scarlet B, Eastone Scarlet BG, Eniacyl Scarlet B, Fenacet Scarlet B, Fluka Disperse Red, Interchem Acetate Scarlet B, Interchem Hisperse Scarlet BH, Intrasperse Scarlet BRN, Kayalon Fast Scarlet B, Microsetile Scarlet B, Miketon Fast Scarlet B, Monocel Scarlet B, N-Ethyl-N-(2-hydroxyethyl)-4-(4-nitrophenylazo)aniline, N-Ethyl-N-(2-hydroxyethyl)-4-[(p-nitrophenyl)azo]aniline, N-Ethyl-N-[p-(p-nitrophenylazo)phenyl]ethanolamine, NSC 57019, Nacelan Scarlet CSB, Neosetile Scarlet B, Nyloquinone Red N, Reliton Scarlet BA, Safaritone Scarlet B, Scarlet Reliton BA, Serinyl Hoisery Scarlet BD, Serisol Fast Scarlet BD, Serisol Fast Scarlet BD 200, Setacyl Scarlet 2BD, Setacyl Scarlet B, Setacyl Scarlet RNA, Silotras Scarlet TSR, Supracet Fast Scarlet B, Tertranese Scarlet N-B, WO 4
InChI Formula InChI=1S/C16H18N4O3/c1-2-19(11-12-21)15-7-3-13(4-8-15)17-18-14-5-9-16(10-6-14)20(22)23/h3-10,21H,2,11-12H2,1H3
SMILES CCN(CCO)c1ccc(cc1)N=Nc2ccc(cc2)[N+](=O)[O-]
IUPAC Name 2-[N-ethyl-4-[(E)-(4-nitrophenyl)diazenyl]anilino]ethanol
Molecular Weight (g/mol) 314.34
Formula Weight 314.1379
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.