missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disperse Red 1 (Technical Grade), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Disperse Red 1 |
| CAS | 2872-52-8 |
| Molecular Formula | C16 H18 N4 O3 |
| Quantity | 500 mg |
| Synonym | C.I. Disperse Red 1 (7CI,8CI), Celliton Scarlet B (6CI), Ethanol, 2-[ethyl[4-[(4-nitrophenyl)azo]phenyl]amino]- (9CI), 2-[Ethyl[4-[2-(4-nitrophenyl)diazenyl]phenyl]amino]ethanol, 1-[N-Ethyl-N-(2-hydroxyethyl)amino]-4-(4-nitrophenylazo)benzene, 4-Nitro-4'-[N-ethyl-N-(β-hydroxyethyl)amino]azobenzene, 4-Nitro-4'-[ethyl(2-hydroxyethyl)amino]azobenzene, 4-[(2-Hydroxyethyl)ethylamino]-4'-nitroazobenzene, 4-[4-(Nitrophenyl)azo]-N-ethyl-N-(2-hydroxyethyl)aniline, 4-[Ethyl(2-hydroxyethyl)amino]-4'-nitroazobenzene, 4-[N-(2-Hydroxyethyl)-N-ethylamino]-4'-nitroazobenzene, 4-[N-Ethyl-N-(2-hydroxyethyl)amino]-4'-nitroazobenzene, Acetamine Scarlet B, Acetate Fast Scarlet B, Acetoquinone Light Scarlet BLZ, Amacel Scarlet GB, C.I. 11110, C.I. Solvent Red 14, Calcosyn Brilliant Scarlet BN, Celliton Discharge Scarlet B, Celliton Scarlet BA-CF, Celutate Scarlet BH, Cibacet Scarlet 2B, Cibacet Scarlet BRN, Cibacet Scarlet BS, Cilla Scarlet B, D 3284, Diacelliton Fast Scarlet B, Diacelliton Scarlet B, Disperse Red 1, Disperse Red 1-glycerol-TDI copolymer, Disperse Red Zh, Disperse Scarlet B, Disperse Scarlet Zh, Dispersive ruby Zh, Dispersol Fast Scarlet B, Dispersol Scarlet B, Durgacet Scarlet B, Eastone Scarlet BG, Eniacyl Scarlet B, Fenacet Scarlet B, Fluka Disperse Red, Interchem Acetate Scarlet B, Interchem Hisperse Scarlet BH, Intrasperse Scarlet BRN, Kayalon Fast Scarlet B, Microsetile Scarlet B, Miketon Fast Scarlet B, Monocel Scarlet B, N-Ethyl-N-(2-hydroxyethyl)-4-(4-nitrophenylazo)aniline, N-Ethyl-N-(2-hydroxyethyl)-4-[(p-nitrophenyl)azo]aniline, N-Ethyl-N-[p-(p-nitrophenylazo)phenyl]ethanolamine, NSC 57019, Nacelan Scarlet CSB, Neosetile Scarlet B, Nyloquinone Red N, Reliton Scarlet BA, Safaritone Scarlet B, Scarlet Reliton BA, Serinyl Hoisery Scarlet BD, Serisol Fast Scarlet BD, Serisol Fast Scarlet BD 200, Setacyl Scarlet 2BD, Setacyl Scarlet B, Setacyl Scarlet RNA, Silotras Scarlet TSR, Supracet Fast Scarlet B, Tertranese Scarlet N-B, WO 4 |
| InChI Formula | InChI=1S/C16H18N4O3/c1-2-19(11-12-21)15-7-3-13(4-8-15)17-18-14-5-9-16(10-6-14)20(22)23/h3-10,21H,2,11-12H2,1H3 |
| SMILES | CCN(CCO)c1ccc(cc1)N=Nc2ccc(cc2)[N+](=O)[O-] |
| IUPAC Name | 2-[N-ethyl-4-[(E)-(4-nitrophenyl)diazenyl]anilino]ethanol |
| Molecular Weight (g/mol) | 314.34 |
| Formula Weight | 314.1379 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction