missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disperse Orange 37, TRC
Specifications
Specifications
| Chemical Name or Material | Disperse Orange 37 |
| CAS | 13301-61-6 |
| Molecular Formula | C17 H15 Cl2 N5 O2 |
| Quantity | 250 mg |
| Synonym | Propanenitrile, 3-[[4-[(2,6-dichloro-4-nitrophenyl)azo]phenyl]ethylamino]- (9CI), Propionitrile, 3-[p-[(2,6-dichloro-4-nitrophenyl)azo]-N-ethylanilino]- (6CI,8CI), 3-[[4-[2-(2,6-Dichloro-4-nitrophenyl)diazenyl]phenyl]ethylamino]propanenitrile, Akasperse Orange RL 150, C.I. Disperse Orange 37, C.I. Disperse Orange 59, C.I. Disperse Orange 76, Disperse Orange 37, Disperse Orange 59, Disperse Orange 76, Disperse Polyester Yellow Brown, Disperse Yellow Brown 2RCW, Eastman Polyester Dark Orange RL, Eastone Dark Orange R-GLF, Intrasil Dark Orange 3GH, Kayalon Polyester Yellow Brown 3RL, Serilene Yellow Brown 2RL, Serilene Yellow Brown 2RL150 |
| InChI Formula | InChI=1S/C17H15Cl2N5O2/c1-2-23(9-3-8-20)13-6-4-12(5-7-13)21-22-17-15(18)10-14(24(25)26)11-16(17)19/h4-7,10-11H,2-3,9H2,1H3 |
| SMILES | CCN(CCC#N)c1ccc(cc1)N=Nc2c(Cl)cc(cc2Cl)[N+](=O)[O-] |
| IUPAC Name | 3-[4-[(E)-(2,6-dichloro-4-nitrophenyl)diazenyl]-N-ethylanilino]propanenitrile |
| Molecular Weight (g/mol) | 392.24 |
| Formula Weight | 391.0603 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?