missing translation for 'onlineSavingsMsg'
Learn More

Disperse Orange 3 (Technical Grade), TRC

Product Code. 30384860
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384860 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384860 Supplier Toronto Research Chemicals Supplier No. TRCD4934855G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Orange 3
CAS 730-40-5
Molecular Formula C12 H10 N4 O2
Quantity 5 g
Synonym Benzenamine, 4-[(4-nitrophenyl)azo]- (9CI), C.I. Disperse Orange 3 (6CI,7CI,8CI), 4-[2-(4-Nitrophenyl)diazenyl]benzenamine, 4-(4-Nitrophenylazo)aniline, 4-(4'-Nitrophenylazo)aniline, 4-Amino-4'-nitroazobenzene, 4-Nitro-4'-aminoazobenzene, 4-[(4-Nitrophenyl)azo]benzenamine, 4'-Nitro-4-aminoazobenzene, Acetamine Orange GR, Acetate Fast Orange G, Acetate Fast Orange GR, Acetoquinone Light Orange JRL, Akasperse Orange RBA, Altocyl Orange GR, Amacel Orange GR, Artisil Direct Orange 2R, Artisil Orange 2R, Atlantic Disperse Orange GRA, C.I. 11005, C.I. Solvent Orange 9, Celliton Discharge Orange GRL, Celliton Fast Orange GR, Celliton Orange GR, Celliton Orange GRA, Celutate Orange GRH, Cibacet Orange 2R, Cibacete Orange 2RN, Cilla Fast Orange GR, Diacelliton Fast Orange GL, Disperse Orange 3, Disperse Orange GR, Disperse Orange Zh, Dispersol Fast Orange G, Dispersol Orange AG, Durgacet Orange GR, Eastman Orange GRN, Eastone Orange 2R, Eniacyl Orange GR, Fenacet Orange G, Interchem Acetate Orange GL, Kayalon Fast Orange GR, Microsetile Orange GR, Miketon Fast Orange GR, Nacelan Orange GR, Neosetile Orange GR, Nyloquinone Orange 2R, Oracet Orange 2R, Ostacet Orange P 2R, Perliton Orange 6R, Reliton Orange GR, Serilic Orange GXD, Setacyl Orange GR, Setacyl Orange RE, Silotras Orange TSGR, Supracet Fast Orange G, Synten Orange 2R, Transetile Orange P-GR, Vonteryl Orange GS, p-(p-Nitrophenylazo)aniline, p-Nitro-p'-aminoazobenzene
InChI Formula InChI=1S/C12H10N4O2/c13-9-1-3-10(4-2-9)14-15-11-5-7-12(8-6-11)16(17)18/h1-8H,13H2
SMILES Nc1ccc(cc1)N=Nc2ccc(cc2)[N+](=O)[O-]
IUPAC Name 4-[(E)-(4-nitrophenyl)diazenyl]aniline
Molecular Weight (g/mol) 242.23
Formula Weight 242.0804
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.