missing translation for 'onlineSavingsMsg'
Learn More

Disperse Orange 1 (>85%), TRC

Product Code. 17975045
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17975045 500 mg 500mg
17904215 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17975045 Supplier Toronto Research Chemicals Supplier No. TRCD495333500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2581-69-3
Molecular Formula C18 H14 N4 O2
Molecular Weight (g/mol) 318.33
Synonym Benzenamine, 4-[(4-nitrophenyl)azo]-N-phenyl- (9CI), C.I. Disperse Orange 1 (6CI,7CI,8CI), 4-[2-(4-Nitrophenyl)diazenyl]-N-phenylbenzenamine, 4-(4-Nitrophenylazo)diphenylamine, 4-(p-Nitrophenylazo)diphenylamine, 4-Anilino-4'-nitroazobenzene, 4-Nitro-4'-(phenylamino)azobenzene, Acetate Orange 5R, Acetoquinone Light Orange 4RL, Akasperse Transfer Orange, C.I. 11080, Celliton Discharge Orange 5RL, Celliton Fast Orange 5R, Cilla Fast Orange 5R, Diacelliton Fast Orange R, Diacelliton Fast Orange RM/D, Disperse Orange 1, Disperse Orange 5R, Dispersol Fast Orange A, Dispersol Orange A, Dispersol Orange B-A, Dispersol Printing Orange A, Miketon Fast Orange 5R, Miketon Polyester Orange 5R, Navilene Orange 5R, Palanil Orange 5R, Reliton Orange 5R, Resolin Orange 5R, Serilene Orange 5R, Serilene Orange 5R300, Serisol Orange 5R, Setacyl Scarlet 2GN, Setacyl Scarlet P 2G, Supracet Scarlet 2G, Vonteryl Red 4G
IUPAC Name 4-[(E)-(4-nitrophenyl)diazenyl]-N-phenylaniline
SMILES [O-][N+](=O)c1ccc(cc1)N=Nc2ccc(Nc3ccccc3)cc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H14N4O2/c23-22(24)18-12-10-17(11-13-18)21-20-16-8-6-15(7-9-16)19-14-4-2-1-3-5-14/h1-13,19H
Formula Weight 318.1117
Chemical Name or Material Disperse Orange 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.