missing translation for 'onlineSavingsMsg'
Learn More

Disperse Diazo Black 3BF, TRC

Product Code. 30381030
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381030 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381030 Supplier Toronto Research Chemicals Supplier No. TRCD493633250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Black 2
CAS 6232-57-1
Molecular Formula C14 H16 N4 O
Quantity 250 mg
Synonym Aniline, 2-methoxy-5-methyl-4,4'-azodi- (7CI), Benzenamine, 4-[(4-aminophenyl)azo]-2-methoxy-5-methyl- (9CI), 4-[2-(4-Aminophenyl)diazenyl]-2-methoxy-5-methylbenzenamine, 1-(4-Aminophenylazo)-2-methyl-4-amino-5-methoxybenzene, 4,4'-Diamino-2-methyl-5-methoxyazobenzene, 4,4'-Diamino-2'-methyl-5'-methoxy-1,1'-azobenzene, Acetamine Diazo Black 3B, Acetoquinone Diazo Navy 2BZ, Amacel Developed Black 3BS, C.I. 11255, C.I. Disperse Black 2, Cibacet Diazo Black B, Diazacyl Black B, Disperse Diazo Black, Disperse Diazo Black 3BF, Dispersol Black 2B, Dispersol Diazo Black 2BN, Fast Black C Base, Kayalon Diazo Black BBS, Meisei Acemyl Diazo Black 3BF, Meisei Teryl Diazo Navy CN, Meisei Teryl Diazo Navy H3B, Naphtoelan Black D Base, Novalon Diazo Black B, Nyloquinone Diazo Navy 2B, Serisol Diazo Black 2B, Setacyl Diazo Black B, Sumikaron Diazo Navy 3B, Terasil Diazo Black B
InChI Formula InChI=1S/C14H16N4O/c1-9-7-12(16)14(19-2)8-13(9)18-17-11-5-3-10(15)4-6-11/h3-8H,15-16H2,1-2H3/b18-17+
SMILES COc1cc(N=Nc2ccc(N)cc2)c(C)cc1N
IUPAC Name 4-[(E)-(4-aminophenyl)diazenyl]-2-methoxy-5-methylaniline
Molecular Weight (g/mol) 256.3
Formula Weight 256.1324
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.