missing translation for 'onlineSavingsMsg'
Learn More

Disperse Blue 60 (Technical Grade), TRC

Product Code. 30382037
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382037 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382037 Supplier Toronto Research Chemicals Supplier No. TRCD493558500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Blue 60
CAS 12217-80-0
Molecular Formula C20H17N3O5
Quantity 500 mg
Synonym 4,11-Diamino-2-(3-methoxypropyl)-1H-naphth[2,3-f]isoindole-1,3,5,10(2H)-tetrone, 2,3-Anthracenedicarboximide, 1,4-diamino-9,10-dihydro-N-(3-methoxypropyl)-9,10-dioxo- (7CI), 2,3-Anthraquinonedicarboximide, 1,4-diamino-N-(3-methoxypropyl)- (6CI), 4,11-Diamino-2-(3-methoxypropyl)-1H-naphth[2,3-f]isoindole-1,3,5,10(2H)-tetrone (ACI), C.I. Disperse Blue 60 (8CI), B 60, C.I. 61104, C.I. Disperse Blue 99, Dianix Turquoise Blue S-BG, Disperse Blue 60, Disperse Blue 99, Disperse Turquoise Blue 60, Disperse Turquoise Blue GL, Disperse Turquoise Blue S-GL, Genacron Brilliant Blue 8G, Kayalon Polyester Turquoise Blue GGF, Kayalon Polyester Turquoise Blue GL-S, Kayalon Polyester Turquoise Blue GL-S 200%, Kayaset B 766, Latyl Brilliant Blue BG, Miketon Polyester Brilliant Blue BG, Palanil Brilliant Blue BG, Palanil Brilliant Blue BG-M, Polycron Turquoise Blue GLP, Qualsperse Blue BNS, Resolin Brilliant Blue BGL, Resolin Brilliant Blue BGL 200, Samaron Blue FFBL, Samaron Brilliant Blue R-GL, Serilene Brilliant Blue 2GN, Sodyecron Brilliant Blue BGE 200, Sumikaron Turquoise Blue G, Sumikaron Turquoise Blue S-GL, Sumikaron Turquoise Blue S-GL 200, Sumikaron Turquoise Blue S-GL double liquid, Sumikaron Turquoise Blue S-GLF, Sumiplast Turquoise Blue G, Synten Blue P-BGFN, Terasil Blue BGE, Terasil Blue BGE 01, Terasil Blue BGE 01-200, Terasil BR Blue BGE 200-017, Terasil Brilliant Blue BGE, Teratop Blue BGE, Tersetile Turquoise GL, TS Turquoise Blue 606, Turquoise Blue S-GL
InChI Formula InChI=1S/C20H17N3O5/c1-28-8-4-7-23-19(26)13-14(20(23)27)16(22)12-11(15(13)21)17(24)9-5-2-3-6-10(9)18(12)25/h2-3,5-6H,4,7-8,21-22H2,1H3
SMILES O=C1C=2C=CC=CC2C(=O)C=3C(N)=C4C(=O)N(C(=O)C4=C(N)C13)CCCOC
IUPAC Name 4,11-Diamino-2-(3-methoxypropyl)-1H-naphth[2,3-f]isoindole-1,3,5,10(2H)-tetrone
Molecular Weight (g/mol) 379.37
Formula Weight 379.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.