missing translation for 'onlineSavingsMsg'
Learn More

Disperse Blue 124, TRC

Product Code. 30382977
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382977 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382977 Supplier Toronto Research Chemicals Supplier No. TRCD49350510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Blue 124
CAS 15141-18-1
Molecular Formula C16 H19 N5 O4 S
Quantity 10 mg
Synonym Ethanol, 2-[N-ethyl-4-[(5-nitro-2-thiazolyl)azo]-m-toluidino]-, acetate (ester) (8CI), Ethanol, 2-[ethyl[3-methyl-4-[(5-nitro-2-thiazolyl)azo]phenyl]amino]-, acetate (ester) (9CI), C.I. 111938, C.I. Disperse Blue 124, Disperse Blue 124, Serisol 3RD, Ethanol, 2-[ethyl[3-methyl-4-[2-(5-nitro-2-thiazolyl)diazenyl]phenyl]amino]-, 1-acetate
InChI Formula InChI=1S/C16H19N5O4S/c1-4-20(7-8-25-12(3)22)13-5-6-14(11(2)9-13)18-19-16-17-10-15(26-16)21(23)24/h5-6,9-10H,4,7-8H2,1-3H3
SMILES CCN(CCOC(=O)C)c1ccc(N=Nc2ncc(s2)[N+](=O)[O-])c(C)c1
IUPAC Name 2-[N-ethyl-3-methyl-4-[(E)-(5-nitro-1,3-thiazol-2-yl)diazenyl]anilino]ethyl acetate
Molecular Weight (g/mol) 377.42
Formula Weight 377.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.