missing translation for 'onlineSavingsMsg'
Learn More

Disperse Blue 106, TRC

Product Code. 30385425
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385425 1 mg
30375396 10 mg
30387503 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385425 Supplier Toronto Research Chemicals Supplier No. TRCD4934701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68516-81-4
Molecular Formula C14 H17 N5 O3 S
Molecular Weight (g/mol) 335.38
Synonym Ethanol, 2-[ethyl[3-methyl-4-[(5-nitro-2-thiazolyl)azo]phenyl]amino]- (9CI), C.I. 111935, C.I. Disperse Blue 106, C.I. Disperse Blue 357, Disperse Blue 106, Disperse Blue 357, Disperse Blue RD, Miketon Polyester Discharge Blue R, Serisol RD 400, Suncron Blue RD 400, Tersetile Blue CRL
IUPAC Name 2-[N-ethyl-3-methyl-4-[(E)-(5-nitro-1,3-thiazol-2-yl)diazenyl]anilino]ethanol
SMILES CCN(CCO)c1ccc(N=Nc2ncc(s2)[N+](=O)[O-])c(C)c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H17N5O3S/c1-3-18(6-7-20)11-4-5-12(10(2)8-11)16-17-14-15-9-13(23-14)19(21)22/h4-5,8-9,20H,3,6-7H2,1-2H3
Formula Weight 335.1052
Chemical Name or Material Disperse Blue 106
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.