Learn More
Fisherbrand™ Dispensing Tip
Product Code. 15541295
Click to view available options
Material:
Glass
Polypropylene
Unit Size:
Each
Product Code. 15541295
Brand: Fisherbrand™ 7822660
Only null left
Add to basket
Add to basket
Featuring Luer lock.
  • Available in glass and polypropylene
  • With Luer Lock
Specifications
Description With Luer lock
Material Glass
For Use With (Application) For 3/16 in. and 1/4 in. ID tubing
Product Type Dispensing Tip