Learn More
Miele Professional™ Dispensing Monitor for Installation PLW8636 APLW 886
Product Code. 30533223 Shop All Miele Products
Click to view available options
Product Type:
Installation Kit
Unit Size:
1 sheet
Product Code. 30533223
Brand: Miele Professional™ 12426910
Monitoring by oval-wheel counter for PLW 8636
  • More precise quantity measurement using oval-wheel counters
  • Monitoring by oval-wheel counter for PLW 8636
  • Installed by Customer Service
TRUSTED_SUSTAINABILITY
Specifications
Product Type Installation Kit
For Use With (Equipment) Washer-Disinfectors
Quantity 1 Ea.