Learn More
Miele Professional™ Dosing system
Shop All Miele Products
Click to view available options
:
for 5 liter canister; integrated level indicator
Description
- Dispenser monitoring – in accordance with DIN EN ISO 15883
- For dispensing liquid cleaning agent or neutralising agent
- For dispensing chemical disinfectant
- 200 mm siphon for 5 litre containers
Specifications
Specifications
| Product Type | Dispensing System |
| For Use With (Equipment) | Washer-disinfectors, laboratory |