Learn More
Poulten Graf™ Dispenser adapter 101.091-38 PTFE white GL38 Optifix
Product Code. 10208260
Product Code. 10208260
Brand: Poulten Graf™ 101 09138
Only null left
Add to basket
Add to basket
For strong acids (excl. HF) and aggressive chemicals. Visible dispensing process, dispensing piston made of PTFE-sleeved, solid glass, with additional sealing lip at the end of the piston to prevent acid fumes and odours from escaping.