missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Dispase (B. polymyxa)

Product Code. 16131840
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16131840

Brand: Abnova™ P5318.1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P29148
For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 36kDa
Name Dispase (B. polymyxa)
Preparation Method Native protein purified from Bacillus polymyxa
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name DISPASE
Assays Neutral Protease (Dispase) Assay; Tissue disaggregation and subcultivation; Prevention of unwanted cell clumping; Preparation of cells for culture; Separation of intact epidermis from dermis and intact epithelial sheet in culture from the substratum; Harvest and transfer of normal, diploid cells and cell lines; Gentle and intact detachment of epidermal cells.
Biological Activity ≥0.1μ/mg dry weight. One Unit releases Folin positive amino acids equivalent to 1 umole tyrosine per minute from casein at 37°C, pH 7.5.
Species Bacteria
Protein Tag None
Expression System Native protein purified from Bacillus polymyxa
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.