Learn More
STEMCELL Technologies Dispase, ACF
Product Code. 17157581
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
17157581 50 mg 50mg
1 options
Product Code. 17157581 Supplier STEMCELL Technologies Supplier No. 1000396
Animal component-free non-specific protease
  • Dispase, Animal Component-Free (ACF) is a neutral, amino-endoprotease that cleaves the N-terminal peptide bond of non-polar amino acid residues and is obtained from cultures free of animal-derived materials
  • Dispase has mild proteolytic activity, which makes it especially useful for the isolation and passaging of primary cells
  • Its proteolytic activity also allows it to maintain membrane integrity
  • Dispase is commonly used with other proteases such as collagenase in cell isolation and tissue dissociation for neural tissue (Dietrich et al.), kidney (Presnell et al.), epithelial tissue (Smoot et al.), endothelial tissue (Müller et al.), lung (Barkauskas et al.), colon (Roig et al.), and stem cells (Thomson et al.; Salmon et al.)
Specifications
Cell Type Chondrocytes, Neural Stem and Progenitor Cells, Osteoblasts
Quantity 50 mg