missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Dispase (1 U/mL)

Product Code. 15569185
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15569185 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15569185 Supplier STEMCELL Technologies Supplier No. 07923

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dispase 1 U/mL in DMEM/F-12

  • Dispase is a protease that is suitable for the gentle dissociation of a wide variety of tissues
  • Incubation of minced tissue with pre-warmed dispase and gentle agitation will liberate cells with minimal cell damage
  • Pre-warmed dispase can also be used to harvest cells from tissue culture plastic
  • Unlike trypsin, dispase is not inhibited by serum
  • Dispase activity is inhibited by EDTA and EGTA
  • Dispase should be removed from cell suspensions by centrifuging the cells followed by washing the cells with buffer or culture medium

Specifications

Cell Type Intestinal Cells, Mammary Cells, Other, Pluripotent Stem Cells, Prostate Cells
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.