missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DISP2 Polyclonal Antibody

Product Code. 12828782
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828782 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828782 Supplier Invitrogen™ Supplier No. PA520631

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20631 can be used with blocking peptide PEP-0751.

DISP2, also known as dispatched homolog 2, is a protein that plays a critical role in the regulation of the hedgehog signaling pathway. It is involved in the transport and release of hedgehog ligands, which are important for cellular differentiation and tissue development. Mutations in the DISP2 gene have been associated with various developmental disorders, including holoprosencephaly and cleft lip and palate. DISP2 is also being studied as a potential therapeutic target in cancer, as the hedgehog pathway is frequently dysregulated in various types of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DISP2
Gene Accession No. A7MBM2, Q8CIP5
Gene Alias AI840597; B230210L08Rik; C15orf36; DISP2; dispatched B; dispatched homolog 2; dispatched homolog 2 (Drosophila); dispatched RND transporter family member 2; DISPB; HsT16908; KIAA1742; LINC00594; mKIAA1742; Protein dispatched homolog 2
Gene Symbols DISP2
Host Species Rabbit
Immunogen A 19 amino acid peptide from near the carboxy terminus of human Disp2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214240, 311324, 85455
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.