missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DISP2 Polyclonal antibody specifically detects DISP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | DISP2 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Dispatched B, Dispatched Homolog 2 (Drosophila), DISPB, HsT16908, KIAA1742, Protein Dispatched Homolog 2 |
| Host Species | Rabbit |
| Immunogen | Disp2 antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human Disp2. The immunogen is located within amino acids 1210 - 1260 of Disp2. Amino Acid Squence: CPALQTSSPYKQAGPS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?