missing translation for 'onlineSavingsMsg'
Learn More

DISP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18403596 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403596 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403596 Supplier Novus Biologicals Supplier No. NBP281818AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DISP1 Polyclonal antibody specifically detects DISP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias DISPAFLJ43740, dispatched A, dispatched homolog 1 (Drosophila), DKFZp434I0428, MGC104180, MGC13130, MGC16796, protein dispatched homolog 1
Host Species Rabbit
Immunogen DISP1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human DISP1. The immunogen is located within amino acids 20 - 70 of DISP1. Amino Acid Squence: CQLTPKEATRTKVSPNG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 84976
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.