missing translation for 'onlineSavingsMsg'
Learn More

Disopyramide Phosphate, TRC

Product Code. 30385706
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385706 500 mg 500mg
30368885 1 g 1g
30377302 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385706 Supplier Toronto Research Chemicals Supplier No. TRCD493455500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22059-60-5
Molecular Formula C21 H29 N3 O . H3 O4 P
Molecular Weight (g/mol) 437.47
Synonym Disopyramide phosphate, (2RS)-4-[bis(1-methylethyl)amino]-2-phenyl-2-(pyridin-2-yl)butanamide dihydrogen phosphate
IUPAC Name 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide;phosphoric acid
SMILES CC(C)N(CCC(C(=O)N)(c1ccccc1)c2ccccn2)C(C)C.OP(=O)(O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H29N3O.H3O4P/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19;1-5(2,3)4/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25);(H3,1,2,3,4)
Formula Weight 437.208
Chemical Name or Material Disopyramide phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.