missing translation for 'onlineSavingsMsg'
Learn More

Disopyramide N-Oxide Dihydrochloride, Mikromol™

Product Code. 30049746
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049746

Brand: Mikromol MM0108.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Disopyramide N-Oxide Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disopyramide N-Oxide Dihydrochloride
CAS 2731709-13-8
Analyte or Component Names Disopyramide N-Oxide Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H29 N3 O2 . 2 Cl H
Certifications/Compliance ISO 9001
Synonym (2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, Disopyramide N-Oxide Dihydrochloride
InChI Formula InChI=1S/C25H31NO3.C2H2O4/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3;3-1(4)2(5)6/h8-10,12-16H,4-7,11,17-18H2,1-3H3;(H,3,4)(H,5,6)
SMILES Cl.Cl.CC(C)[N+]([O-])(CCC(C(=O)N)(c1ccccc1)c2ccccn2)C(C)C
IUPAC Name 4-amino-4-oxo-3-phenyl-N,N-di(propan-2-yl)-3-pyridin-2-ylbutan-1-amine oxide;dihydrochloride
Molecular Weight (g/mol) 428.4
Quantity 25 mg
Formula Weight 427.1793 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.