missing translation for 'onlineSavingsMsg'
Learn More

Disodium Tetrachloropalladate, TRC

Product Code. 30376850
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376850 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376850 Supplier Toronto Research Chemicals Supplier No. TRCD4933355G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disodium Tetrachloropalladate
CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Quantity 5 g
Synonym Disodium Palladium Tetrachloride, Disodium Tetrachloropalladate(2-), Disodium Tetrachloropalladium, Palladium Sodium Chloride (PdNa2Cl4), Sodium Chloropalladate, Sodium Chloropalladate, Sodium Palladium Chloride, Sodium Palladium Tetrachloride, Sodium Tetrachloropalladate, Sodium Tetrachloropalladate(2-)
InChI Formula InChI=1S/4ClH.2Na.Pd/h4*1H;;;/q;;;;2*+1;+2/p-4
SMILES [Na+].[Na+].Cl[Pd-2](Cl)(Cl)Cl
IUPAC Name disodium;tetrachloropalladium(2-)
Molecular Weight (g/mol) 294.21
Formula Weight 291.7584
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.