missing translation for 'onlineSavingsMsg'
Learn More

Disodium Tellurite, TRC

Product Code. 30420699
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420699 10 g 10g
30412052 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30420699 Supplier Toronto Research Chemicals Supplier No. TRCS67363010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-20-2
Molecular Formula 2 Na . O3 Te
Molecular Weight (g/mol) 221.58
Synonym Telluric acid (H2TeO3), sodium salt (1:2), Sodium tellurate(IV) (Na2TeO3) (6CI,7CI), Telluric acid (H2TeO3), disodium salt (8CI,9CI), Disodium tellurate (Na2TeO3), Disodium tellurite, Disodium trioxotellurate, Sodium tellurate (Na2TeO3), Sodium tellurite, Sodium tellurite (Na2TeO3), Sodium tellurium oxide (Na2TeO3)
IUPAC Name disodium;tellurite
SMILES [Na+].[Na+].[O-][Te](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/2Na.H2O3Te/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2
Formula Weight 223.8705
Chemical Name or Material Disodium Tellurite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.