missing translation for 'onlineSavingsMsg'
Learn More

Disodium Anthraquinone-2,6-disulfonate, TRC

Product Code. 30385109
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385109 250 mg 250mg
30369468 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385109 Supplier Toronto Research Chemicals Supplier No. TRCD493648250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 853-68-9
Molecular Formula C14H6Na2O8S2
Molecular Weight (g/mol) 412.29
Synonym AQS, Anthraquinone-2,6-disulfonic Acid Disodium Salt, Disodium 9,10-Anthraquinone-2,6-disulfonate, Disodium 9,10-Dioxoanthracene-2,6-disulfonate, Disodium Anthraquinone-2,6-disulfonate, Sodium Anthraquinone-2,6-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)c1ccc2C(=O)c3cc(ccc3C(=O)c2c1)S(=O)(=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H8O8S2.2Na/c15-13-9-3-1-7(23(17,18)19)5-11(9)14(16)10-4-2-8(6-12(10)13)24(20,21)22;;/h1-6H,(H,17,18,19)(H,20,21,22);;/q;2*+1/p-2
Formula Weight 411.9299
Chemical Name or Material Disodium Anthraquinone-2,6-disulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.