missing translation for 'onlineSavingsMsg'
Learn More

Disodium 3-Hydroxy-4-[(4-methyl-2-sulfonatophenyl)diazenyl]-2-naphthoate, TRC

Product Code. 30384151
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384151 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384151 Supplier Toronto Research Chemicals Supplier No. TRCD4950031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disodium 3-Hydroxy-4-[(4-methyl-2-sulfonatophenyl)diazenyl]-2-naphthoate
CAS 5858-81-1
Molecular Formula C18 H12 N2 O6 S . 2 Na
Quantity 1 g
Synonym 3-Hydroxy-4-[2-(4-methyl-2-sulfophenyl)diazenyl]-2-naphthalenecarboxylic Acid Sodium Salt, 11070 Red, C 19-6619, C.I. 15850, CI 15850 red, D and C Red 6, D and C Red No. 6, D&C Red 6, D&C Red No. 6, FD and C Red No. 6, FD&C Red No. 6, Fuji Carmine 6B, Fuji Carmine 6B660, Irgalite Red 4B, Irgalite Rubine KB 4N, Irgalite Rubine PB, Isol Bona Rubine BK, Isol Bona Rubine BKS, Isol Bona Rubine KBK, Isol Ruby BK, Isol Ruby BKS, Japan Red 201, Japan Red No. 201, Kromon Permanent Red 4B, LC 303, Lithol Rubine B, Lithol Rubine BNA, Lithol Rubine Red, Toner DK, PR 57, Permanent Red 4B, Permanent Red F 6R, Pigment Red 57, Pigment Rubine B, Pigment Rubine BCL, Plastol Rubine BC, Red 201, Red 6, Red No. 201, Resamine Rubine BC, Rubine Red RR 1253, Segnale Red 3R, Silotermo Carmine G, Siloton Rubine 2B, Siloton Rubine B, Unipure Red LC 303, Vynamon Claret Y, ZAB 120
InChI Formula InChI=1S/C18H14N2O6S.2Na/c1-10-6-7-14(15(8-10)27(24,25)26)19-20-16-12-5-3-2-4-11(12)9-13(17(16)21)18(22)23;;/h2-9,21H,1H3,(H,22,23)(H,24,25,26);;/q;2*+1/p-2
SMILES [Na+].[Na+].Cc1ccc(N=Nc2c(O)c(cc3ccccc23)C(=O)[O-])c(c1)S(=O)(=O)[O-]
IUPAC Name disodium;3-hydroxy-4-[(E)-(4-methyl-2-sulfonatophenyl)diazenyl]naphthalene-2-carboxylate
Molecular Weight (g/mol) 430.342
Formula Weight 430.021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.