Learn More
Johnson Johnson Medical™ Disinfectant granules Presept
Product Code. 12378667
Encompass_Preferred
Product Code. 12378667
Brand: Johnson Johnson Medical™ SPG500
Reacts with water in body fluid spillages to produce a rapid acting wide spectrum disinfectant effective against bacteria, fungi, viruses (including AIDS virus) and bacterial spores.
Effervescent, containing sodium dichloroisocyanurate (NaDCC). The granules are applied over the spillage, left for two minutes and removed for disposal. The surface is then wiped with a damp cloth, which is disposed of along with used granules. Should not be mixed with acids or cationic detergents.Use biocides safely. Always read the label and product information before use.
Granules contain and solidify the spillage, enabling easy removal, and preventing the spread of contamination.
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.