Learn More
Tego™ Disinfectant 2000, ClearKlens Tego™
Product Code. 12834991
Change view
Click to view available options
Volume (Metric):
5 L
Product Code. Volume (Metric)
12834991 5 L
1 options
Product Code. 12834991 Supplier Tego™ Supplier No. 7509458
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Tego™ 2000 is a versatile disinfectant that is used to clean and disinfect in sterile environments. It has an active amphoteric-base with a broad spectrum of antimicrobial activity, it is suitable for open plant cleaning applications throughout the food, beverage and dairy industry.
  • Ready to use surface disinfectant
  • Amphoteric-based, antimicrobial
  • For use in sterile environments
Specifications
Volume (Metric) 5 L