missing translation for 'onlineSavingsMsg'
Learn More

Dishevelled-3 Antibody, Novus Biologicals™

Product Code. 18327275 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327275 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327275 Supplier Novus Biologicals Supplier No. NBP303068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dishevelled-3 Polyclonal specifically detects Dishevelled-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dishevelled-3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias dishevelled 3 (homologous to Drosophila dsh), dishevelled, dsh homolog 3 (Drosophila), Dishevelled-3, DSH homolog 3, KIAA0208dishevelled-3, segment polarity protein dishevelled homolog DVL-3
Host Species Rabbit
Immunogen A synthetic peptide of human Dishevelled-3.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1857
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.